missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-propylphosphonate, 97%, Thermo Scientific Chemicals

Catalog No. p-7023499
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1710509 10 g
AAA1710518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1710509 Supplier Thermo Scientific Chemicals Supplier No. A1710509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diethyl 1-propylphosphonate, is used as a medical intermediate and in organic synthesis. It is also used in electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1-propylphosphonate, is used as a medical intermediate and in organic synthesis. It is also used in electronics industry.

Solubility
Soluble in water (27 g/L) (25°C).

Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to oxidizing agents, reducing agents, acids, bases.

Chemical Identifiers

CAS 18812-51-6
Molecular Formula C7H17O3P
Molecular Weight (g/mol) 180.184
MDL Number MFCD00015139
InChI Key RUIKOPXSCCGLOM-UHFFFAOYSA-N
Synonym diethyl propylphosphonate, diethyl propanephosphonate, diethyl 1-propylphosphonate, phosphonic acid, propyl-, diethyl ester, diethyl 1-propanephosphonate, diethyl-n-propylphosphonate, diethoxypropylphosphine oxide, acmc-20al9i, diethyl n-propylphosphonate, 1-diethoxyphosphoryl-propane
PubChem CID 140428
IUPAC Name 1-diethoxyphosphorylpropane
SMILES CCCP(=O)(OCC)OCC

Specifications

Boiling Point 94°C to 96°C (12 mmHg)
Refractive Index 1.419
Quantity 10 g
Beilstein 1762076
Solubility Information Soluble in water (27g/L) (25°C).
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material Diethyl 1-propylphosphonate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only