missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,4-Phenylenediacrylate, TCI America™

Catalog No. P13455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13455G Supplier TCI America Supplier No. P13455G
Only null left

Chemical Identifiers

CAS 17088-28-7
Molecular Formula C16H18O4
Molecular Weight (g/mol) 274.316
MDL Number MFCD00017521
InChI Key QYGWZBFQWUBYAT-WGDLNXRISA-N
Synonym 1,4-Phenylenediacrylic Acid Diethyl Ester
PubChem CID 6441554
IUPAC Name ethyl (E)-3-[4-[(E)-3-ethoxy-3-oxoprop-1-enyl]phenyl]prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)C=CC(=O)OCC

Specifications

Melting Point 95°C
Color Yellow
Quantity 5 g
Formula Weight 274.32
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 1,4-Phenylenediacrylate