missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,3-Acetonedicarboxylate 95.0+%, TCI America™

Catalog No. p-7116684
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
A005525ML 25 mL
A0055500ML 500 mL
2 options

Catalog No. A005525ML

Supplier: TCI America A005525ML

Only null left

Chemical Identifiers

CAS 105-50-0
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00009200
InChI Key ZSANYRMTSBBUCA-UHFFFAOYSA-N
Synonym diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester
PubChem CID 66045
IUPAC Name 1,5-diethyl 3-oxopentanedioate
SMILES CCOC(=O)CC(=O)CC(=O)OCC

Specifications

Color Yellow
Boiling Point 250°C
Quantity 25 mL
Formula Weight 202.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 1,3-Acetonedicarboxylate