missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 1,2-Cyclohexanedicarboxylate 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 135°C |
| Quantity | 25 g |
| Formula Weight | 228.29 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl 1,2-Cyclohexanedicarboxylate |
Safety and Handling
| EINECSNumber | (3)-2435 |
| RTECSNumber | GU9275000 |
| TSCA | No |