missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,2-Cyclohexanedicarboxylate 95.0+%, TCI America™

Catalog No. C0476500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0476500G 500 g
C047625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0476500G Supplier TCI America Supplier No. C0476500G
Only null left

Chemical Identifiers

CAS 10138-59-7
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.288
MDL Number MFCD00029449
InChI Key ZTUZDYWYNQDJKR-UHFFFAOYSA-N
Synonym 1,2-Cyclohexanedicarboxylic Acid Diethyl Ester
PubChem CID 24990
IUPAC Name diethyl cyclohexane-1,2-dicarboxylate
SMILES CCOC(=O)C1CCCCC1C(=O)OCC

Specifications

Color Colorless
Boiling Point 135°C
Quantity 500 g
Formula Weight 228.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 1,2-Cyclohexanedicarboxylate
EINECSNumber (3)-2435
RTECSNumber GU9275000
TSCA No