missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,1-Cyclopropanedicarboxylate 96.0+%, TCI America™

Catalog No. p-7124925
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13415G 5 g
C134125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13415G Supplier TCI America Supplier No. C13415G
Only null left

Chemical Identifiers

CAS 1559-02-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00001283
InChI Key KYYUCZOHNYSLFV-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclopropanedicarboxylate, 1,1-diethyl cyclopropane-1,1-dicarboxylate, 1,1-cyclopropanedicarboxylic acid diethyl ester, 1,1-cyclopropanedicarboxylic acid, diethyl ester, cyclopropane-1,1-dicarboxylic acid diethyl ester, 1,1-bis ethoxycarbonyl cyclopropane, diethylcyclopropan-1,1-dicarboxylat, diethyl-1,1-cyclopropanedicarboxylate, acmc-209dcw, ksc495e2n
PubChem CID 73790
IUPAC Name 1,1-diethyl cyclopropane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CC1)C(=O)OCC

Specifications

Color Colorless
Boiling Point 215°C
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 1,1-Cyclopropanedicarboxylate