missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,1-cyclobutanedicarboxylate, 95%

Catalog No. p-7026237
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2214009 10 g
AAB2214018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2214009 Supplier Thermo Scientific Chemicals Supplier No. B2214009

CAS: 3779-29-1 | C10H16O4 | 200.23 g/mol

Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3779-29-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00019261
InChI Key JPNJEJSZSMXWSV-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclobutanedicarboxylate, 1,1-cyclobutanedicarboxylic acid, diethyl ester, 1,1-diethyl cyclobutane-1,1-dicarboxylate, cyclobutane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid, 1,1-diethyl ester, 1,1-dicarboethoxycyclobutane, ethyl 1-ethoxycarbonyl cyclobutanecarboxylate, acmc-1cmcx, diethyl1,1-cyclobutanedicarboxylate
PubChem CID 77410
IUPAC Name diethyl cyclobutane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CCC1)C(=O)OCC

Specifications

Density 1.05
Boiling Point 96°C to 98°C (8 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.436
Quantity 10 g
Beilstein 2050195
Solubility Information Insoluble in water.
Formula Weight 200.24
Percent Purity 95%
Chemical Name or Material Diethyl 1,1-cyclobutanedicarboxylate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 223-239-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only