missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylvinylsilane 97.0+%, TCI America™

Catalog No. D231825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D231825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D231825ML Supplier TCI America Supplier No. D231825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5507-44-8
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009067
InChI Key MBGQQKKTDDNCSG-UHFFFAOYSA-N
Synonym diethoxy methyl vinyl silane, methylvinyldiethoxysilane, silane, ethenyldiethoxymethyl, vinylmethyldiethoxysilane, diethoxy methyl vinylsilane, silane, diethoxymethylvinyl, vinyldiethoxymethylsilane, vinylmethyldiathoxysilan, diethoxymethylvinyl silane, diethoxy-methyl-vinyl-silane
PubChem CID 79646
IUPAC Name ethenyl-diethoxy-methylsilane
SMILES CCO[Si](C)(C=C)OCC

Specifications

Color Yellow
Boiling Point 134°C
Quantity 25 mL
UN Number 1993
Formula Weight 160.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethoxymethylvinylsilane
EINECSNumber (2)-2055
TSCA Yes