missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylvinylsilane, 97%

Catalog No. p-7046409
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1422414 25 g
AAL1422422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1422414 Supplier Thermo Scientific Chemicals Supplier No. L1422414
Only null left

CAS: 5507-44-8 | C7H16O2Si | 160.288 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5507-44-8
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009067
InChI Key MBGQQKKTDDNCSG-UHFFFAOYSA-N
Synonym diethoxy methyl vinyl silane, methylvinyldiethoxysilane, silane, ethenyldiethoxymethyl, vinylmethyldiethoxysilane, diethoxy methyl vinylsilane, silane, diethoxymethylvinyl, vinyldiethoxymethylsilane, vinylmethyldiathoxysilan, diethoxymethylvinyl silane, diethoxy-methyl-vinyl-silane
PubChem CID 79646
IUPAC Name ethenyl-diethoxy-methylsilane
SMILES CCO[Si](C)(C=C)OCC

Specifications

Boiling Point 133°C to 134°C
Quantity 25 g
UN Number UN1993
Beilstein 970124
Flash Point 17°C (62°F)
Formula Weight 160.29
Percent Purity 97%
Refractive Index 1.4
Sensitivity Moisture sensitive
Density 0.858
Chemical Name or Material Diethoxymethylvinylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 226-850-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only