missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylphenylsilane, 97+%

Catalog No. p-7027150
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2368409 10 g
AAB2368418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2368409 Supplier Thermo Scientific Chemicals Supplier No. B2368409
Only null left

CAS: 775-56-4 | C11H18O2Si | 210.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 775-56-4
Molecular Formula C11H18O2Si
Molecular Weight (g/mol) 210.35
MDL Number MFCD00048541
InChI Key MNFGEHQPOWJJBH-UHFFFAOYSA-N
Synonym methylphenyldiethoxysilane, diethoxy methyl phenylsilane, diethoxy methyl phenyl silane, silane, diethoxymethylphenyl, benzene, diethoxymethylsilyl, pubchem10811, acmc-209pap, methyl-phenyl-diethoxysilane, diethoxy-methyl-phenyl-silane
PubChem CID 69891
IUPAC Name diethoxy-methyl-phenylsilane
SMILES CCO[Si](C)(OCC)C1=CC=CC=C1

Specifications

Boiling Point 117°C to 118°C (30mmHg)
Quantity 10 g
Beilstein 2938396
Flash Point 88°C (190°F)
Formula Weight 210.35
Percent Purity ≥97%
Refractive Index 1.4696
Sensitivity Moisture sensitive
Density 0.963
Chemical Name or Material Diethoxymethylphenylsilane
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-275-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only