missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethoxymethane, 99%, stab. with 100ppm BHT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 66°C |
| Density | 0.831 |
| Boiling Point | 87°C to 88°C |
| Flash Point | -5°C (23°F) |
| Assay Percent Range | 0.99 |
| Refractive Index | 1.374 |
| Quantity | 100 mL |
| UN Number | UN2373 |
| Beilstein | 1697253 |
| Formula Weight | 104.15 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H225-H311-H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H225-H311-H315-H319-H335 |
| missing translation for 'healthHazard3' | P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P372-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIETHOXYMETHANE |
| EINECSNumber | 207-330-6 |
| RTECSNumber | PA8500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only