missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethane, 99%, pure, stabilized

Catalog No. p-107529
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296450050 5 mL Glass bottle
AC296455000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296450050 Supplier Thermo Scientific Chemicals Supplier No. 296450050

CAS: 462-95-3 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 462-95-3 , 128-37-0
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00009246
InChI Key KLKFAASOGCDTDT-UHFFFAOYSA-N
Synonym diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
PubChem CID 10024
IUPAC Name ethoxymethoxyethane
SMILES CCOCOCC

Specifications

Melting Point -66.5°C
Color Colorless
Density 0.8310g/mL
Boiling Point 87.5°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3720 to 1.3740
Linear Formula CH3CH2OCH2OCH2CH3
Quantity 5 mL
Specific Gravity 0.831
Solubility Information Solubility in water: 4.2%. Other solubilities: miscible with ethanol, ethyl ether, very soluble in acetone, soluble in benzene, chloroform
Stabilizer 100ppm BHT max.
Viscosity 0.405 mPa.s (20°C)
Formula Weight 104.15
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Diethoxymethane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume
EINECSNumber 207-330-6
RTECSNumber PA8500000
TSCA TSCA

RUO – Research Use Only