missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydimethylsilane 98.0+%, TCI America™

Catalog No. p-7126221
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D045425ML 25 mL
D0454100ML 100 mL
D0454500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D045425ML Supplier TCI America Supplier No. D045425ML

Chemical Identifiers

CAS 78-62-6
Molecular Formula C6H16O2Si
Molecular Weight (g/mol) 148.277
MDL Number MFCD00009068
InChI Key YYLGKUPAFFKGRQ-UHFFFAOYSA-N
Synonym dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22
PubChem CID 62322
IUPAC Name diethoxy(dimethyl)silane
SMILES CCO[Si](C)(C)OCC

Specifications

Melting Point -70°C
Color Colorless
Boiling Point 114°C
Quantity 25 mL
UN Number 2380
Formula Weight 148.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethoxydimethylsilane
EINECSNumber (2)-2052&(2)-2053
RTECSNumber VV3590000
TSCA Yes