missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydimethylsilane, 97%

Catalog No. AAL1472518
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1472518 50 g
AAL1472530 250 g
AAL147250B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1472518 Supplier Thermo Scientific Chemicals Supplier No. L1472518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 78-62-6 | C6H16O2Si | 148.277 g/mol

Diethoxydimethylsilane is useful for blocking hydroxyl and amino groups in organic synthesis reactions. This silylating step allows subsequent reactions to be carried out which would be adversely affected by the presence of active hydrogen in the hydroxyl or amine groups. Following the reaction step, hydroxyl or amine groups blocked with diethoxydimethylsilane may be recovered by a hydrolysis procedure. It is also used for preparing hydrophobic and release materials as well as enhancing flow of powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethoxydimethylsilane is useful for blocking hydroxyl and amino groups in organic synthesis reactions. This silylating step allows subsequent reactions to be carried out which would be adversely affected by the presence of active hydrogen in the hydroxyl or amine groups. Following the reaction step, hydroxyl or amine groups blocked with diethoxydimethylsilane may be recovered by a hydrolysis procedure. It is also used for preparing hydrophobic and release materials as well as enhancing flow of powders.

Solubility
Reacts with water.

Notes
Store under dry inert gas. It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents

Chemical Identifiers

CAS 78-62-6
Molecular Formula C6H16O2Si
Molecular Weight (g/mol) 148.277
MDL Number MFCD00009068
InChI Key YYLGKUPAFFKGRQ-UHFFFAOYSA-N
Synonym dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22
PubChem CID 62322
IUPAC Name diethoxy(dimethyl)silane
SMILES CCO[Si](C)(C)OCC

Specifications

Boiling Point 111°C to 113°C
Melting Point -70°C
Quantity 50 g
UN Number UN2380
Beilstein 1736110
Flash Point 11°C (52°F)
Solubility Information Reacts with water.
Formula Weight 148.28
Percent Purity 97%
Odor Mild
Refractive Index 1.382
Sensitivity Moisture sensitive
Density 0.865
Chemical Name or Material Diethoxydimethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H335
Highly flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLDIETHOXYSILANE
EINECSNumber 201-127-6
RTECSNumber VV3590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only