missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine Hydrochloride 95.0+%, TCI America™

Supplier:  TCI America I0009500G

 View more versions of this product

Catalog No. I0009500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diethanolamine Hydrochloride
C4H12ClNO2
500 g
VJLOFJZWUDZJBX-UHFFFAOYSA-N
bis(2-hydroxyethyl)azanium chloride
518946
≥95.0% (T,N)
14426-21-2
MFCD00012595
diethanolamine hydrochloride, diethanolamine hcl, ethanol, 2,2'-iminobis-, hydrochloride, unii-oi45ouw092, ethanol, 2,2'-iminobis-, hydrochloride 1:1, bis 2-hydroxyethyl ammonium chloride, acmc-1cf9m, c4h11no2.hcl, diethanolamine hydrochloride salt, diethanolaminehydrochloride
[Cl-].OCC[NH2+]CCO
141.60
141.60
Liquid

Chemical Identifiers

14426-21-2
141.60
VJLOFJZWUDZJBX-UHFFFAOYSA-N
518946
[Cl-].OCC[NH2+]CCO
C4H12ClNO2
MFCD00012595
diethanolamine hydrochloride, diethanolamine hcl, ethanol, 2,2'-iminobis-, hydrochloride, unii-oi45ouw092, ethanol, 2,2'-iminobis-, hydrochloride 1:1, bis 2-hydroxyethyl ammonium chloride, acmc-1cf9m, c4h11no2.hcl, diethanolamine hydrochloride salt, diethanolaminehydrochloride
bis(2-hydroxyethyl)azanium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : KL3500000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions