missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine Hydrochloride 95.0+%, TCI America™

Catalog No. I000925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I000925G 25 g
I0009500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I000925G Supplier TCI America Supplier No. I000925G

Chemical Identifiers

CAS 14426-21-2
Molecular Formula C4H12ClNO2
Molecular Weight (g/mol) 141.60
MDL Number MFCD00012595
InChI Key VJLOFJZWUDZJBX-UHFFFAOYSA-N
Synonym diethanolamine hydrochloride, diethanolamine hcl, ethanol, 2,2'-iminobis-, hydrochloride, unii-oi45ouw092, ethanol, 2,2'-iminobis-, hydrochloride 1:1, bis 2-hydroxyethyl ammonium chloride, acmc-1cf9m, c4h11no2.hcl, diethanolamine hydrochloride salt, diethanolaminehydrochloride
PubChem CID 518946
IUPAC Name bis(2-hydroxyethyl)azanium chloride
SMILES [Cl-].OCC[NH2+]CCO

Specifications

Quantity 25 g
Formula Weight 141.60
Percent Purity ≥95.0% (T,N)
Physical Form Liquid
Chemical Name or Material Diethanolamine Hydrochloride
RTECSNumber KL3500000
TSCA Yes