missing translation for 'onlineSavingsMsg'
Learn More

Didecylmethylamine 95.0+%, TCI America™

Catalog No. D231125ML
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
D231125ML 25 mL
D2311500ML 500 mL
2 options

Catalog No. D231125ML

Supplier: TCI America D231125ML

Only null left

Chemical Identifiers

CAS 7396-58-9
Molecular Formula C21H45N
Molecular Weight (g/mol) 311.598
MDL Number MFCD00077727
InChI Key ATBNMWWDBWBAHM-UHFFFAOYSA-N
PubChem CID 81880
IUPAC Name N-decyl-N-methyldecan-1-amine
SMILES CCCCCCCCCCN(C)CCCCCCCCCC

Specifications

Melting Point -7°C
Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 2735
Formula Weight 311.60
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Didecylmethylamine
EINECSNumber (2)-0176
TSCA Yes