missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Didecyldimethylammonium Bromide 98.0+%, TCI America™
Supplier: TCI America D236325G
Specifications
| Didecyldimethylammonium Bromide | |
| White-Yellow | |
| MFCD00012194 | |
| didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide | |
| CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] | |
| 406.537 | |
| 406.54 | |
| Crystalline Powder |
| 2390-68-3 | |
| C22H48BrN | |
| 25 g | |
| UMGXUWVIJIQANV-UHFFFAOYSA-M | |
| didecyl(dimethyl)azanium;bromide | |
| 16957 | |
| ≥98.0% (T) |
Chemical Identifiers
| 2390-68-3 | |
| 406.537 | |
| UMGXUWVIJIQANV-UHFFFAOYSA-M | |
| 16957 | |
| CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] |
| C22H48BrN | |
| MFCD00012194 | |
| didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide | |
| didecyl(dimethyl)azanium;bromide |
Safety and Handling
EINECSNumber : (2)-0184&(9)-1971
TSCA : No