missing translation for 'onlineSavingsMsg'
Learn More

Didecylamine 97.0+%, TCI America™

Catalog No. p-7126218
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04465G 5 g
D044625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04465G Supplier TCI America Supplier No. D04465G
Only null left

Chemical Identifiers

CAS 1120-49-6
Molecular Formula C20H43N
Molecular Weight (g/mol) 297.571
MDL Number MFCD00048396
InChI Key GMTCPFCMAHMEMT-UHFFFAOYSA-N
Synonym didecylamine, 1-decanamine, n-decyl, di-n-decylamine, bis decyl amine, unii-82r4o8ep59, di c10-c25 alkyl, amines, di-c10-25-alkyl, di-decylamine, didecyl amine
PubChem CID 14261
IUPAC Name N-decyldecan-1-amine
SMILES CCCCCCCCCCNCCCCCCCCCC

Specifications

Color White
Quantity 5 g
Formula Weight 297.57
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Didecylamine
EINECSNumber (2)-0176
TSCA Yes