missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylphosphinyl Chloride 98.0+%, TCI America™

Catalog No. D22055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22055G Supplier TCI America Supplier No. D22055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15873-72-0
Molecular Formula C12H22ClOP
Molecular Weight (g/mol) 248.731
MDL Number MFCD00191344
InChI Key HKYSIUYOULVHTO-UHFFFAOYSA-N
PubChem CID 557982
IUPAC Name [chloro(cyclohexyl)phosphoryl]cyclohexane
SMILES C1CCC(CC1)P(=O)(C2CCCCC2)Cl

Specifications

Melting Point 109°C
Quantity 5 g
UN Number 1759
Formula Weight 248.73
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylphosphinyl Chloride
TSCA No
Recommended Storage Refrigerator