missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylamine 99.0+%, TCI America™

Catalog No. D0435500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0435500ML 500 mL
D043525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0435500ML Supplier TCI America Supplier No. D0435500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-83-7
Molecular Formula C12H23N
Molecular Weight (g/mol) 181.32
MDL Number MFCD00011658
InChI Key XBPCUCUWBYBCDP-UHFFFAOYSA-N
Synonym dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
PubChem CID 7582
ChEBI CHEBI:34694
IUPAC Name N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1

Specifications

Melting Point 0°C
Color Yellow
Boiling Point 256°C
Quantity 500 mL
UN Number 2565
Formula Weight 181.32
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclohexylamine
EINECSNumber (3)-2259&(3)-2686
RTECSNumber HY4025000
TSCA Yes