missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylamine Borane 95.0+%, TCI America™

Catalog No. p-7124113
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55455G 5 g
B554525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55455G Supplier TCI America Supplier No. B55455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 131765-96-3
Molecular Formula C12H26BN
Synonym Borane - Dicyclohexylamine Complex

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 195.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylamine Borane
Recommended Storage Refrigerator