Learn More
Dicyclohexylamine, 98%
CAS: 101-83-7 | C12H23N | 181.32 g/mol
Supplier: Thermo Scientific Chemicals A156710F
Description
Dicyclohexylamine is used as an intermediate for the production of antioxidants and vulcanization accelerator. It is used as a catalyst for flexible polyurethane foams. It is also used in agrochemicals and textile chemicals. Used to constitute ionic liquid matrices for bacterial analysis in matrix assisted laser desorption/ionisation mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDicyclohexylamine is used as an intermediate for the production of antioxidants and vulcanization accelerator. It is used as a catalyst for flexible polyurethane foams. It is also used in agrochemicals and textile chemicals. Used to constitute ionic liquid matrices for bacterial analysis in matrix assisted laser desorption/ionisation mass spectrometry.
Solubility
Sparingly soluble in water (0.8 g/L).
Notes
Air sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container.
Specifications
| Dicyclohexylamine | |
| -2°C | |
| 256°C | |
| C12H23N | |
| MFCD00011658 | |
| UN2565 | |
| Air Sensitive | |
| dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech | |
| XBPCUCUWBYBCDP-UHFFFAOYSA-N | |
| N-cyclohexylcyclohexanamine | |
| 7582 | |
| 181.32 |
| 101-83-7 | |
| 0.913 | |
| 99°C (210°F) | |
| 1.4842 | |
| 2500 mL | |
| 605923 | |
| 14,3095 | |
| Sparingly soluble in water (0.8g/L). | |
| C1CCC(CC1)NC1CCCCC1 | |
| 181.32 | |
| CHEBI:34694 | |
| 98% |
Chemical Identifiers
| 101-83-7 | |
| 181.32 | |
| XBPCUCUWBYBCDP-UHFFFAOYSA-N | |
| 7582 | |
| N-cyclohexylcyclohexanamine |
| C12H23N | |
| MFCD00011658 | |
| dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech | |
| CHEBI:34694 | |
| C1CCC(CC1)NC1CCCCC1 |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: DICYCLOHEXYLAMINE
EINECSNumber : 202-980-7
RTECSNumber : HY4025000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only