missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylacetic acid, 98+%

Catalog No. AAH3352314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3352314 25 g
AAH3352306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3352314 Supplier Thermo Scientific Chemicals Supplier No. H3352314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52034-92-1 | C14H24O2 | 224.344 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 52034-92-1
Molecular Formula C14H24O2
Molecular Weight (g/mol) 224.344
MDL Number MFCD00075017
InChI Key PGGMEZOUAPIYOY-UHFFFAOYSA-N
Synonym dicyclohexylacetic acid, hexahydro-diphenylessig, dicyclohexyl acetic acid, dicyclohexyl-acetic acid, acmc-20ala4
PubChem CID 3661562
IUPAC Name 2,2-dicyclohexylacetic acid
SMILES C1CCC(CC1)C(C2CCCCC2)C(=O)O

Specifications

Melting Point 139°C to 141°C
Quantity 25 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 224.34
Percent Purity ≥98%
Chemical Name or Material Dicyclohexylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only