missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclohexyl(trifluoromethanesulfonyloxy)borane 98.0+%, TCI America™
$126.76 - $391.44
Chemical Identifiers
| CAS | 145412-54-0 |
|---|---|
| Molecular Formula | C13H22BF3O3S |
| Molecular Weight (g/mol) | 326.181 |
| MDL Number | MFCD06797094 |
| InChI Key | FBVDLYBQGQLANQ-UHFFFAOYSA-N |
| Synonym | Dicyclohexylboron Triflate, Trifluoromethanesulfonic Acid Dicyclohexylboryl Ester |
| PubChem CID | 10958346 |
| IUPAC Name | dicyclohexylboranyl trifluoromethanesulfonate |
| SMILES | B(C1CCCCC1)(C2CCCCC2)OS(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 145412-54-0 | |
| 326.181 | |
| FBVDLYBQGQLANQ-UHFFFAOYSA-N | |
| 10958346 | |
| B(C1CCCCC1)(C2CCCCC2)OS(=O)(=O)C(F)(F)F |
| C13H22BF3O3S | |
| MFCD06797094 | |
| Dicyclohexylboron Triflate, Trifluoromethanesulfonic Acid Dicyclohexylboryl Ester | |
| dicyclohexylboranyl trifluoromethanesulfonate |
Specifications
| 145412-54-0 | |
| C13H22BF3O3S | |
| 1 g | |
| Dicyclohexylboron Triflate, Trifluoromethanesulfonic Acid Dicyclohexylboryl Ester | |
| B(C1CCCCC1)(C2CCCCC2)OS(=O)(=O)C(F)(F)F | |
| 326.181 | |
| 326.18 | |
| Crystalline Powder |
| Yellow | |
| MFCD06797094 | |
| 1759 | |
| FBVDLYBQGQLANQ-UHFFFAOYSA-N | |
| dicyclohexylboranyl trifluoromethanesulfonate | |
| 10958346 | |
| ≥97.0% (T) | |
| Dicyclohexyl(trifluoromethanesulfonyloxy)borane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator