missing translation for 'onlineSavingsMsg'
Learn More

Dicyandiamide, 99%

Catalog No. AAA1045130
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1045130 250 g
AAA1045136 500 g
AAA104510E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1045130 Supplier Thermo Scientific Chemicals Supplier No. A1045130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 461-58-5 | C2H4N4 | 84.08 g/mol

Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

Solubility
Soluble in Water (32g/L ) at 20°C

Notes
Store at 2-8°C. Incompatible with strong acids, strong oxidizing agents, strong acids, strong bases, ammonium nitrate, potassium chlorates.

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008066
InChI Key QGBSISYHAICWAH-UHFFFAOYSA-N
Synonym dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
PubChem CID 10005
IUPAC Name 2-cyanoguanidine
SMILES NC(N)=NC#N

Specifications

Melting Point 208°C to 211°C
Density 1.4
Odor Odorless
Assay Percent Range 99%
Quantity 250 g
Beilstein 605637
Merck Index 14,3092
Solubility Information Soluble in Water (32g/L ) at 20°C
Formula Weight 84.08
Percent Purity 99%
Chemical Name or Material Dicyandiamide
Show More Show Less
EINECSNumber 207-312-8
RTECSNumber ME9950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only