missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Dicot Stem Model

Catalog No. S13503
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13503 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13503 Supplier Eisco™ Supplier No. BM0011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The dicot stem model shows the inner anatomy and vasculature of a dicot stem. The cross-section view with a slice removed provides two different internal views of a dicot stem.

Specifications

Color Assorted
For Use With (Application) Understanding the structure and function of a dicot stem
Grade 9 to 12
Includes Key card included with scientific labels for each corresponding number on the model
Material ABS Plastic
Quantity 1
Gender Male, Female
Product Type Dicot Stem Model (Transverse and Longitudinal Section)