missing translation for 'onlineSavingsMsg'
Learn More

Dicloxacillin sodium salt monohydrate

Catalog No. p-7058203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6158103 1 g
AAJ6158106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6158103 Supplier Thermo Scientific Chemicals Supplier No. J6158103
Only null left

CAS: 13412-64-1 | C19H21Cl2N3NaO6S | 513.342 g/mol

Dicloxacillin sodium salt is used in the treatment of staphylococci resistant to penicillin G. Dicloxacillin binds to specific penicillin-binding proteins in the bacterial cell wall and therefore inhibits the last stage of bacterial cell wall synthesis. Cell lysis, mediated by bacterial cell wall autolytic enzymes, is the result. Dicloxacillin may interfere with autolysin inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 13412-64-1
Molecular Formula C19H21Cl2N3NaO6S
Molecular Weight (g/mol) 513.342
MDL Number MFCD00056865
InChI Key ASMFXKXNAWVTAY-MBGFYOAHSA-N
PubChem CID 131673926
IUPAC Name (2S,5R,6R)-6-[[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate
SMILES [HH].CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na]

Specifications

Melting Point 222°C to 225°C
Color White
Quantity 1 g
Merck Index 14,3084
Solubility Information Soluble in water
Formula Weight 492.33
Physical Form Powder
Chemical Name or Material Dicloxacillin sodium salt
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 603-794-2
RTECSNumber XH8925000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only