missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylphosphine 98.0+%, TCI America™

Catalog No. P020725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020725ML 25 mL
P0207500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P020725ML Supplier TCI America Supplier No. P020725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 644-97-3
Molecular Formula C6H5Cl2P
Molecular Weight (g/mol) 178.98
MDL Number MFCD00000528
InChI Key IMDXZWRLUZPMDH-UHFFFAOYSA-N
Synonym dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl
PubChem CID 12573
IUPAC Name dichloro(phenyl)phosphane
SMILES ClP(Cl)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 59°C
Quantity 25 mL
UN Number 2798
Formula Weight 178.98
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Dichlorophenylphosphine
EINECSNumber (3)-2517
RTECSNumber TB2478000
TSCA Yes