Learn More
Dichlorophenylphosphine, 97%
CAS: 644-97-3 | C6H5Cl2P | 178.98 g/mol
$56.49 - $393.66
Chemical Identifiers
| CAS | 644-97-3 |
|---|---|
| Molecular Formula | C6H5Cl2P |
| Molecular Weight (g/mol) | 178.98 |
| MDL Number | MFCD00000528 |
| InChI Key | IMDXZWRLUZPMDH-UHFFFAOYSA-N |
| Synonym | dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl |
| PubChem CID | 12573 |
| IUPAC Name | dichloro(phenyl)phosphane |
| SMILES | ClP(Cl)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1028418
|
Thermo Scientific Chemicals
A1028418 |
50 g |
Each for $56.49
|
|
|||||
|
AAA1028430
|
Thermo Scientific Chemicals
A1028430 |
250 g |
Each for $146.00
|
|
|||||
|
AAA102840B
|
Thermo Scientific Chemicals
A102840B |
1000 g |
Each for $393.66
|
|
|||||
Description
Dichlorophenylphosphine and its derivatives are used as an intermediate to make plasticizers, antioxidants, stabilizers, catalysts and high pressure lubricant additives and in lacquer formulatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDichlorophenylphosphine and its derivatives are used as an intermediate to make plasticizers, antioxidants, stabilizers, catalysts and high pressure lubricant additives and in lacquer formulatings.
Solubility
Reacts with water.
Notes
Air & moisture Sensitive. Incompatible with bases, oxidizing agents, water/ moisture & air. Store under inert gas.
Chemical Identifiers
| 644-97-3 | |
| 178.98 | |
| IMDXZWRLUZPMDH-UHFFFAOYSA-N | |
| 12573 | |
| ClP(Cl)C1=CC=CC=C1 |
| C6H5Cl2P | |
| MFCD00000528 | |
| dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl | |
| dichloro(phenyl)phosphane |
Specifications
| 644-97-3 | |
| 1.319 | |
| 102°C (215°F) | |
| C6H5Cl2P | |
| MFCD00000528 | |
| UN2798 | |
| Air and moisture sensitive | |
| Reacts with water. | |
| ClP(Cl)C1=CC=CC=C1 | |
| 178.98 | |
| 178.99 | |
| Dichlorophenylphosphine |
| -51°C | |
| 99°C to 101°C (11 mmHg) | |
| Pungent | |
| 1.598 | |
| 50 g | |
| 508189 | |
| dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl | |
| IMDXZWRLUZPMDH-UHFFFAOYSA-N | |
| dichloro(phenyl)phosphane | |
| 12573 | |
| 97% |
Safety and Handling
GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P222-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H250-H301-H314-H335
DOTInformation : Hazard Class: 4.2; Packaging Group: I
EINECSNumber : 211-425-8
RTECSNumber : TB2478000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only