missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylborane 98.0+%, TCI America™

Catalog No. D43675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43675G 5 g
D436725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43675G Supplier TCI America Supplier No. D43675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 873-51-8
Molecular Formula C6H5BCl2
Molecular Weight (g/mol) 158.816
MDL Number MFCD00013612
InChI Key NCQDQONETMHUMY-UHFFFAOYSA-N
Synonym phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron
PubChem CID 136678
IUPAC Name dichloro(phenyl)borane
SMILES B(C1=CC=CC=C1)(Cl)Cl

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 177°C
Quantity 5 g
UN Number 2924
Formula Weight 158.82
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Dichlorophenylborane