Learn More
Dichlorophenylborane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 318550250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dichlorophenylborane | |
| 7.0°C | |
| 1.2200g/mL | |
| −14°C | |
| 96% min. (ex Cl) (Argentometry) | |
| C6H5BCl2 | |
| C6H5BCl2 | |
| 25 g | |
| phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron | |
| B(C1=CC=CC=C1)(Cl)Cl | |
| 158.82 | |
| 158.82 | |
| Liquid |
| 873-51-8 | |
| Colorless | |
| 175.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5450 to 1.5470 | |
| MFCD00013612 | |
| 1.22 | |
| NCQDQONETMHUMY-UHFFFAOYSA-N | |
| dichloro(phenyl)borane | |
| 136678 | |
| 97% |
Chemical Identifiers
| 873-51-8 | |
| 158.82 | |
| NCQDQONETMHUMY-UHFFFAOYSA-N | |
| 136678 | |
| B(C1=CC=CC=C1)(Cl)Cl |
| C6H5BCl2 | |
| MFCD00013612 | |
| phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron | |
| dichloro(phenyl)borane |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Recommended Storage : Product darkens on storage
RUO â Research Use Only