Learn More
Dichlorophenylborane, 96%
CAS: 873-51-8 | C6H5BCl2 | 158.816 g/mol
$236.55 - $236.55
Chemical Identifiers
| CAS | 873-51-8 |
|---|---|
| Molecular Formula | C6H5BCl2 |
| Molecular Weight (g/mol) | 158.816 |
| MDL Number | MFCD00013612 |
| InChI Key | NCQDQONETMHUMY-UHFFFAOYSA-N |
| Synonym | phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron |
| PubChem CID | 136678 |
| IUPAC Name | dichloro(phenyl)borane |
| SMILES | B(C1=CC=CC=C1)(Cl)Cl |
Description
Dichlorophenylborane is widely used as a catalyst in Antiproliferative macrolide and cell migration inhibitor lactimidomycin and in enantioselective synthesis of polypeptide segments using vinylogous Mukaiyama aldol reactions and in enantioselective Diels-Alder cycloadditions after its reaction with allo-threonine derivatives. It is a stereo selective alkylative ring opening of cyclic anhydrides and in cross-metathesis reaction of amino derivatives with olefines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 873-51-8 | |
| 158.816 | |
| NCQDQONETMHUMY-UHFFFAOYSA-N | |
| 136678 | |
| B(C1=CC=CC=C1)(Cl)Cl |
| C6H5BCl2 | |
| MFCD00013612 | |
| phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron | |
| dichloro(phenyl)borane |
Specifications
| 873-51-8 | |
| Colorless | |
| 175°C | |
| Acrid | |
| 1.545 | |
| 5 g | |
| 2243576 | |
| phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron | |
| NCQDQONETMHUMY-UHFFFAOYSA-N | |
| dichloro(phenyl)borane | |
| 136678 | |
| 96% | |
| Dichlorophenylborane |
| 7°C | |
| 1.194 g/mL | |
| 14°C (57°F) | |
| C6H5BCl2 | |
| MFCD00013612 | |
| UN2924 | |
| Moisture sensitive | |
| Soluble in water (Reacts violently). | |
| B(C1=CC=CC=C1)(Cl)Cl | |
| 158.816 | |
| 158.83 | |
| Liquid |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only