missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylphenylsilane, 98%

Catalog No. p-7027328
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2399914 25 g
AAB2399936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2399914 Supplier Thermo Scientific Chemicals Supplier No. B2399914
Only null left

CAS: 149-74-6 | C7H8Cl2Si | 191.13 g/mol

Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is sensitive to moisture. Protect from humidity and water.

Chemical Identifiers

CAS 149-74-6
Molecular Formula C7H8Cl2Si
Molecular Weight (g/mol) 191.13
MDL Number MFCD00000490
InChI Key GNEPOXWQWFSSOU-UHFFFAOYSA-N
Synonym methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
PubChem CID 9006
IUPAC Name dichloro-methyl-phenylsilane
SMILES C[Si](Cl)(Cl)C1=CC=CC=C1

Specifications

Boiling Point 205°C to 206°C
Quantity 25 g
Linear Formula C6H5Si(CH3)Cl2
UN Number UN2437
Beilstein 970975
Flash Point 82°C (179°F)
Solubility Information Reacts with water.
Formula Weight 191.13
Percent Purity 98%
Odor Acrid
Refractive Index 1.519
Sensitivity Moisture sensitive
Density 1.173
Chemical Name or Material Dichloromethylphenylsilane
Show More Show Less
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHYLPHENYLDICHLOROSILANE
EINECSNumber 205-746-2
RTECSNumber VV3530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only