missing translation for 'onlineSavingsMsg'
Learn More

Dichloroisocyanuric acid sodium salt monohydrate, 97%

Catalog No. p-4258711
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436450500 50 g Glass bottle
AC436452500 250 g Glass bottle
AC436450010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC436450500 Supplier Thermo Scientific Chemicals Supplier No. 436450500
Only null left

CAS: 52671-45-1 | C3Cl2N3NaO3·H2O | 237.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 52671-45-1
Molecular Formula C3Cl2N3NaO3·H2O
Molecular Weight (g/mol) 237.96
InChI Key FIHQYHSNTSJNGG-UHFFFAOYSA-N
Synonym Sodium dichloroisocyanurate dihydrate
PubChem CID 129893375
IUPAC Name 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium;dihydrate
SMILES C1(=O)NC(=O)N(C(=O)N1Cl)Cl.O.O.[Na]

Specifications

Melting Point 240.0°C to 250.0°C
Color White
Physical Form Crystals or Granules
Quantity 50 g
Assay Percent Range 96.0 to 104% (on dry substance) (Iodometry)
Solubility Information Solubility in water: ca 250g/L (25°C)
Formula Weight 237.96
Percent Purity 97%
Infrared Spectrum Authentic
Loss on Drying 15% max.
Packaging Glass bottle
Chemical Name or Material Dichloroisocyanuric acid sodium salt monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates t
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

RUO – Research Use Only