missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloroisocyanuric acid sodium salt monohydrate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Melting Point | 240.0°C to 250.0°C |
| Color | White |
| Physical Form | Crystals or Granules |
| Quantity | 1 kg |
| Assay Percent Range | 96.0 to 104% (on dry substance) (Iodometry) |
| Solubility Information | Solubility in water: ca 250g/L (25°C) |
| Formula Weight | 237.96 |
| Percent Purity | 97% |
| Infrared Spectrum | Authentic |
| Loss on Drying | 15% max. |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. Very toxic to aquatic life with long lasting effects. Contact with acids liberates t |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major |
RUO – Research Use Only