missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiphenylsilane 98.0+%, TCI America™

Catalog No. D036225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D036225G 25 g
D0362100G 100 g
D0362500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D036225G Supplier TCI America Supplier No. D036225G

Chemical Identifiers

CAS 80-10-4
Molecular Formula C12H10Cl2Si
Molecular Weight (g/mol) 253.20
MDL Number MFCD00000489
InChI Key OSXYHAQZDCICNX-UHFFFAOYSA-N
Synonym diphenyldichlorosilane, silane, dichlorodiphenyl, dichloro diphenyl silane, diphenylsilicon dichloride, diphenylsilyl dichloride, dichlor-difenylsilan, diphenyl dichlorosilane, dichlor-difenylsilan czech, hsdb 316, benzene, 1,1'-dichlorosilylene bis
PubChem CID 6627
IUPAC Name dichlorodiphenylsilane
SMILES Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 303°C
Quantity 25 g
UN Number 1769
Formula Weight 253.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorodiphenylsilane
EINECSNumber (3)-2634
RTECSNumber VV3190000
TSCA Yes