missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylsilane, 99+%

CAS: 75-78-5 | C2H6Cl2Si | 129.06 g/mol

Supplier:  Thermo Scientific Chemicals 113310010

 View more versions of this product

Catalog No. AC113310010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dichlorodimethylsilane
-76.0°C
1.0600g/mL
−9°C
99% min. (GC)
C2H6Cl2Si
(CH3)2SiCl2
03,114; 04,183; 17,113
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dichloro(dimethyl)silane
6398
129.06
Liquid
75-78-5
Colorless to Yellow
70.0°C
Authentic
Glass bottle
1.4030 to 1.4050
1 L
1.06
Solubility in water: reacts. Other solubilities: soluble in proton-free solvents (care must be,taken with ketones and esters),soluble in benzene and ether,decomposes in ethanol
C[Si](C)(Cl)Cl
129.06
0.6 mPa.s (25°C)
99+%

Chemical Identifiers

75-78-5
129.06
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
dichloro(dimethyl)silane
C2H6Cl2Si
LIKFHECYJZWXFJ-UHFFFAOYSA-N
6398
C[Si](C)(Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 200-901-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only