missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylsilane, 99+%, AcroSeal™

Catalog No. p-4229172
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430881000 100 mL AcroSeal™ Glass Bottle
AC430888000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430881000 Supplier Thermo Scientific Chemicals Supplier No. 430881000
Only null left

CAS: 75-78-5 | C2H6Cl2Si | 129.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-78-5
Molecular Formula C2H6Cl2Si
Molecular Weight (g/mol) 129.06
MDL Number MFCD00000491
InChI Key LIKFHECYJZWXFJ-UHFFFAOYSA-N
Synonym dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
PubChem CID 6398
IUPAC Name dichloro(dimethyl)silane
SMILES C[Si](C)(Cl)Cl

Specifications

Melting Point -76.0°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 70.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4030 to 1.4050
Linear Formula (CH3)2SiCl2
Quantity 100 mL
Specific Gravity 1.06
Solubility Information Solubility in water: reacts. Other solubilities: soluble in proton-free solvents (care must be,taken with ketones and esters),soluble in benzene and ether,decomposes in ethanol
Viscosity 0.6 mPa.s (25°C)
Formula Weight 129.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dichlorodimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 200-901-

RUO – Research Use Only