missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylsilane, 98+%

CAS: 75-78-5 | C2H6Cl2Si | 129.055 g/mol

Supplier:  Thermo Scientific Chemicals L121330B

 View more versions of this product

Catalog No. AAL121330B


Only null left
Explore available promotions

Description

Description

Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with alcohols, amines and strong bases.
Specifications

Specifications

70°C to 71°C
∼-76°C
1000 g
(CH3)2SiCl2
UN1162
−5°C (23°F)
Immiscible with water.
C[Si](C)(Cl)Cl
129.055
129.06
1.4045
1.068
Dichlorodimethylsilane
75-78-5
C2H6Cl2Si
MFCD00000491
605287
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dichloro(dimethyl)silane
6398
99%
Moisture sensitive

Chemical Identifiers

75-78-5
129.055
LIKFHECYJZWXFJ-UHFFFAOYSA-N
6398
C[Si](C)(Cl)Cl
C2H6Cl2Si
MFCD00000491
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
dichloro(dimethyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H331-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLDICHLOROSILANE

EINECSNumber : 200-901-0

RTECSNumber : VV3150000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only