missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiethylsilane, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1719-53-5
Molecular Formula C4H10Cl2Si
Molecular Weight (g/mol) 157.11
MDL Number MFCD00000493
InChI Key BYLOHCRAPOSXLY-UHFFFAOYSA-N
Synonym diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
PubChem CID 15589
IUPAC Name dichloro(diethyl)silane
SMILES CC[Si](Cl)(Cl)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC338100050
SDS View Documents
Thermo Scientific Chemicals
338100050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC338100250
SDS View Documents
Thermo Scientific Chemicals
338100250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1719-53-5
157.11
BYLOHCRAPOSXLY-UHFFFAOYSA-N
15589
CC[Si](Cl)(Cl)CC
C4H10Cl2Si
MFCD00000493
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
dichloro(diethyl)silane

Specifications

1719-53-5
128.0°C to 129.0°C
96% min. (GC)
C4H10Cl2Si
MFCD00000493
1.05
Solubility in water: reacts
CC[Si](Cl)(Cl)CC
157.11
157.12
Dichlorodiethylsilane, 97%
1.0500g/mL
28°C
Glass bottle
(CH3CH2)2Si(Cl)2
5g
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
BYLOHCRAPOSXLY-UHFFFAOYSA-N
dichloro(diethyl)silane
15589
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 217-005-0

RTECSNumber : VV3060000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only