missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiethylsilane, 94%

CAS: 1719-53-5 | C4H10Cl2Si | 157.11 g/mol

Supplier:  Thermo Scientific Chemicals L0323909

 View more versions of this product

Catalog No. AAL0323909


Only null left
Explore available promotions

Description

Description

Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorodiethylsilane is used in ethchlorvynol assay. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water and ether.

Notes
Moisture Sensitive. Store at 4°C. Protect from moisture.Incompatible materials are Oxidizing agents, Bases, Water.
Specifications

Specifications

129°C to 131°C
-97°C
10 g
MFCD00000493
605313
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
BYLOHCRAPOSXLY-UHFFFAOYSA-N
dichloro(diethyl)silane
15589
94%
1.431
1.055
Dichlorodiethylsilane
1719-53-5
C4H10Cl2Si
UN1767
28°C (82°F)
Slightly soluble in water and ether.
CC[Si](Cl)(Cl)CC
157.11
157.12
Acrid
Moisture sensitive

Chemical Identifiers

1719-53-5
157.11
BYLOHCRAPOSXLY-UHFFFAOYSA-N
15589
CC[Si](Cl)(Cl)CC
C4H10Cl2Si
MFCD00000493
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
dichloro(diethyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIETHYLDICHLOROSILANE

EINECSNumber : 217-005-0

RTECSNumber : VV3060000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only