missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodecylmethylsilane 97.0+%, TCI America™

Catalog No. D234825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D234825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D234825ML Supplier TCI America Supplier No. D234825ML
Only null left

Chemical Identifiers

CAS 18051-88-2
Molecular Formula C11H24Cl2Si
Molecular Weight (g/mol) 255.30
MDL Number MFCD00053194
InChI Key UFVFNJHZBMHRCO-UHFFFAOYSA-N
Synonym Decylmethyldichlorosilane
PubChem CID 87434
IUPAC Name dichloro(decyl)methylsilane
SMILES CCCCCCCCCC[Si](C)(Cl)Cl

Specifications

Color Yellow
Boiling Point 129°C
Quantity 25 mL
UN Number 2987
Formula Weight 255.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorodecylmethylsilane
EINECSNumber (2)-2041
TSCA Yes