missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II), Pd 15%

Catalog No. AA4551103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4551103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4551103 Supplier Thermo Scientific Chemicals Supplier No. 4551103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 887919-35-9 | C32H56Cl2N2P2Pd | 708.08 g/mol

Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) is a highly active, air-stable catalyst for Suzuki-Miyaura cross-coupling with aryl halides including 5- and 6-membered heteroaryl chlorides.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 887919-35-9
Molecular Formula C32H56Cl2N2P2Pd
Molecular Weight (g/mol) 708.08
MDL Number MFCD09265123
InChI Key DWOZNANUEDYIOF-UHFFFAOYSA-L
Synonym bis di-tert-butyl 4-dimethylaminophenyl phosphine dichloropalladium ii, pd amphos cl2, pdcl2 amphos 2, a-taphos 2pdcl2, dichlorobis di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii, alpha-taphos 2pdcl2, bis 4-di-tert-butylphosphino-n,n-dimethylbenzenamine palladium dichloride
PubChem CID 11714597
IUPAC Name 4-ditert-butylphosphanyl-N,N-dimethylaniline;dichloropalladium
SMILES Cl[Pd]Cl.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C

Specifications

Color Yellow
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 708.08
Physical Form Crystals
Chemical Name or Material Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only