missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(2-ethylhexyl)silane 95.0+%, TCI America™

Supplier:  TCI America D44435G

 View more versions of this product

Catalog No. D44435G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dichlorobis(2-ethylhexyl)silane
Yellow
MFCD22370170
2987
CCCCC(CC)C[Si](CC(CC)CCCC)(Cl)Cl
325.433
325.43
Liquid
1089687-03-5
C16H34Cl2Si
5 g
BXDGSXMFTGZLDT-UHFFFAOYSA-N
dichloro-bis(2-ethylhexyl)silane
22023834
≥95.0% (GC)

Chemical Identifiers

1089687-03-5
325.433
BXDGSXMFTGZLDT-UHFFFAOYSA-N
dichloro-bis(2-ethylhexyl)silane
C16H34Cl2Si
MFCD22370170
22023834
CCCCC(CC)C[Si](CC(CC)CCCC)(Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions