missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(2-ethylhexyl)silane 95.0+%, TCI America™

Catalog No. D44431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44431G 1 g
D44435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44431G Supplier TCI America Supplier No. D44431G
Only null left

Chemical Identifiers

CAS 1089687-03-5
Molecular Formula C16H34Cl2Si
Molecular Weight (g/mol) 325.433
MDL Number MFCD22370170
InChI Key BXDGSXMFTGZLDT-UHFFFAOYSA-N
PubChem CID 22023834
IUPAC Name dichloro-bis(2-ethylhexyl)silane
SMILES CCCCC(CC)C[Si](CC(CC)CCCC)(Cl)Cl

Specifications

Color Yellow
Quantity 1 g
UN Number 2987
Formula Weight 325.43
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorobis(2-ethylhexyl)silane