missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloroacetyl Chloride 98.0+%, TCI America™
$98.79 - $324.35
Chemical Identifiers
| CAS | 79-36-7 |
|---|---|
| Molecular Formula | C2HCl3O |
| Molecular Weight (g/mol) | 147.379 |
| MDL Number | MFCD00000840 |
| InChI Key | FBCCMZVIWNDFMO-UHFFFAOYSA-N |
| Synonym | dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove |
| PubChem CID | 6593 |
| ChEBI | CHEBI:34688 |
| IUPAC Name | 2,2-dichloroacetyl chloride |
| SMILES | C(C(=O)Cl)(Cl)Cl |
Chemical Identifiers
| 79-36-7 | |
| 147.379 | |
| FBCCMZVIWNDFMO-UHFFFAOYSA-N | |
| 6593 | |
| 2,2-dichloroacetyl chloride |
| C2HCl3O | |
| MFCD00000840 | |
| dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove | |
| CHEBI:34688 | |
| C(C(=O)Cl)(Cl)Cl |
Specifications
| 79-36-7 | |
| 108°C | |
| MFCD00000840 | |
| 1765 | |
| FBCCMZVIWNDFMO-UHFFFAOYSA-N | |
| 2,2-dichloroacetyl chloride | |
| 6593 | |
| 147.38 | |
| Liquid |
| Yellow | |
| C2HCl3O | |
| 25 g | |
| dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove | |
| C(C(=O)Cl)(Cl)Cl | |
| 147.379 | |
| CHEBI:34688 | |
| ≥98.0% (GC,T) | |
| Dichloroacetyl Chloride |
Safety and Handling
EINECSNumber : (2)-1162
RTECSNumber : AO6650000
TSCA : Yes