missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetyl Chloride 98.0+%, TCI America™

Catalog No. D031325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D031325G 25 g
D0313500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D031325G Supplier TCI America Supplier No. D031325G

Chemical Identifiers

CAS 79-36-7
Molecular Formula C2HCl3O
Molecular Weight (g/mol) 147.379
MDL Number MFCD00000840
InChI Key FBCCMZVIWNDFMO-UHFFFAOYSA-N
Synonym dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
PubChem CID 6593
ChEBI CHEBI:34688
IUPAC Name 2,2-dichloroacetyl chloride
SMILES C(C(=O)Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
UN Number 1765
Formula Weight 147.38
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Dichloroacetyl Chloride
EINECSNumber (2)-1162
RTECSNumber AO6650000
TSCA Yes