missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetyl chloride, 97%

Catalog No. AAB2229622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2229622 100 g
AAB2229636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2229622 Supplier Thermo Scientific Chemicals Supplier No. B2229622

CAS: 79-36-7 | C2HCl3O | 147.379 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-36-7
Molecular Formula C2HCl3O
Molecular Weight (g/mol) 147.379
MDL Number MFCD00000840
InChI Key FBCCMZVIWNDFMO-UHFFFAOYSA-N
Synonym dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
PubChem CID 6593
ChEBI CHEBI:34688
IUPAC Name 2,2-dichloroacetyl chloride
SMILES C(C(=O)Cl)(Cl)Cl

Specifications

Density 1.532
Boiling Point 107°C to 108°C
Flash Point None
Refractive Index 1.462
Quantity 100 g
UN Number UN1765
Sensitivity Moisture sensitive
Merck Index 14,3053
Formula Weight 147.39
Percent Purity 97%
Chemical Name or Material Dichloroacetyl chloride
Show More Show Less
Hazard Category H227-H311-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETYL CHLORIDE
EINECSNumber 201-199-9
RTECSNumber AO6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only