missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer

CAS: 12354-84-6 | C20H30Cl4Ir2 | 796.694 g/mol

$365.42 - $365.42

Chemical Identifiers

CAS 12354-84-6
Molecular Formula C20H30Cl4Ir2
Molecular Weight (g/mol) 796.694
MDL Number MFCD00075435
InChI Key MMAGMBCAIFVRGJ-UHFFFAOYSA-J
Synonym unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di
PubChem CID 76030743
IUPAC Name iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride
SMILES C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45565ME
View Documents
Thermo Scientific Chemicals
045565ME
500 mg
Each for $365.42
Only null left
 
Description

Description

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer is used as a precursor to catalysts for the asymmetric transfer hydrogenation of ketones. It is a catalyst for greener amine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12354-84-6
796.694
MMAGMBCAIFVRGJ-UHFFFAOYSA-J
76030743
C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3]
C20H30Cl4Ir2
MFCD00075435
unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di
iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride

Specifications

12354-84-6
MFCD00075435
unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di
C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3]
796.694
796.67
C20H30Cl4Ir2
500 mg
MMAGMBCAIFVRGJ-UHFFFAOYSA-J
iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride
76030743
Dichloro (pentamethylcyclopentadienyl)iridium (III) dimer
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only